Assay Details
| Gene Symbol: IFI16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319449.1 | NP_001306378.1 | 7 - 8 | 1499 | 70 | |
| XM_005541319.2 | XP_005541376.1 | 8 - 9 | 1792 | 70 | ||
| GenBank mRNA | KF154419.1 | - | 7 - 8 | 1499 | 70 | |
More Information
Set Membership: |
|

IFI16
Genomic Map