Assay Details
| Gene Symbol: PYHIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541320.2 | XP_005541377.1 | 3 - 4 | 511 | 113 | |
| XM_015455114.1 | XP_015310600.1 | 2 - 3 | 1092 | 113 | ||
More Information
Set Membership: |
|

PYHIN1
Genomic Map