Assay Details
| Gene Symbol: FCRLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541149.2 | XP_005541206.1 | 5 - 6 | 1088 | 96 | |
| XM_005541150.2 | XP_005541207.1 | 4 - 5 | 1070 | 96 | ||
More Information
Set Membership: |
|

FCRLA
Genomic Map