Assay Details
| Gene Symbol: MRPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285365.1 | NP_001272294.1 | 6 - 7 | 787 | 78 | |
| GenBank mRNA | AB170181.1 | - | 6 - 7 | 787 | 78 | |
More Information
Set Membership: |
|

MRPL3
Genomic Map