Assay Details
| Gene Symbol: PCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548264.2 | XP_005548321.1 | 1 - 2 | 186 | 140 | |
| GenBank mRNA | AB169544.1 | - | 1 - 2 | 106 | 140 | |
More Information
Set Membership: |
|

PCNP
Genomic Map