Assay Details
| Gene Symbol: SMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593159.2 | XP_005593216.1 | 10 - 11 | 1308 | 136 | |
| XM_005593161.2 | XP_005593218.1 | 10 - 11 | 1157 | 136 | ||
| XM_015443588.1 | XP_015299074.1 | 10 - 11 | 1138 | 136 | ||
More Information
Set Membership: |
|

SMS
Genomic Map