Assay Details
| Gene Symbol: ACKR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574718.2 | XP_005574775.1 | 2 | 3040 | 169 | |
| XM_005574719.2 | XP_005574776.1 | 2 | 1823 | 169 | ||
| XM_005574720.2 | XP_005574777.1 | 2 | 2228 | 169 | ||
| XM_015433015.1 | XP_015288501.1 | 3 | 1989 | 169 | ||
| XM_015433016.1 | XP_015288502.1 | 2 | 1894 | 169 | ||
More Information
Set Membership: |
|

ACKR3
Genomic Map