Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591151.2 | XP_005591208.1 | 2 - 3 | 708 | 95 | |
| XM_015442457.1 | XP_015297943.1 | 2 - 3 | 620 | 95 | ||
| XM_015442458.1 | XP_015297944.1 | 2 - 3 | 556 | 95 | ||
More Information
Set Membership: |
|

ZNF500
Genomic Map