Assay Details
| Gene Symbol: RTP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546909.2 | XP_005546966.1 | 1 - 2 | 1369 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RTP3| Gene Symbol: RTP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546909.2 | XP_005546966.1 | 1 - 2 | 1369 | 128 | |
Set Membership: |
|