Assay Details
| Gene Symbol: TMEM191C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567880.2 | XP_005567937.1 | 4 - 5 | 823 | 134 | |
| XM_005567882.2 | XP_005567939.1 | 4 - 5 | 822 | 134 | ||
| XM_005567883.2 | XP_005567940.1 | 4 - 5 | 821 | 134 | ||
| XM_015457395.1 | XP_015312881.1 | 3 - 4 | 772 | 134 | ||
More Information
Set Membership: |
|

TMEM191C
Genomic Map