Assay Details
| Gene Symbol: GPR174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594012.2 | XP_005594069.1 | 3 | 1967 | 108 | |
| XM_005594013.2 | XP_005594070.1 | 3 | 7065 | 108 | ||
More Information
Set Membership: |
|

GPR174
Genomic Map