Assay Details
| Gene Symbol: MAFG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585280.2 | XP_005585337.1 | 2 - 3 | 3593 | 69 | |
| XM_015437962.1 | XP_015293448.1 | 2 - 3 | 3072 | 69 | ||
More Information
Set Membership: |
|

MAFG
Genomic Map