Assay Details
| Gene Symbol: RWDD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283328.1 | NP_001270257.1 | 2 - 3 | 407 | 141 | |
| XM_015448828.1 | XP_015304314.1 | 2 - 3 | 631 | 141 | ||
| GenBank mRNA | AB169140.1 | - | 2 - 3 | 407 | 141 | |
More Information
Set Membership: |
|

RWDD2A
Genomic Map