Assay Details
| Gene Symbol: IP6K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553281.2 | XP_005553338.1 | 5 - 6 | 1180 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IP6K3| Gene Symbol: IP6K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553281.2 | XP_005553338.1 | 5 - 6 | 1180 | 99 | |
Set Membership: |
|