Assay Details
| Gene Symbol: TMEM217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553131.2 | XP_005553188.2 | 2 | 628 | 112 | |
| XM_015449466.1 | XP_015304952.1 | 2 | 632 | 112 | ||
| XM_015449467.1 | XP_015304953.1 | 3 | 584 | 112 | ||
More Information
Set Membership: |
|

TMEM217
Genomic Map