Assay Details
| Gene Symbol: ACSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443196.1 | XP_015298682.1 | 2 - 3 | 843 | 119 | |
| XM_015443197.1 | XP_015298683.1 | 2 - 3 | 1360 | 119 | ||
| XM_015443199.1 | XP_015298685.1 | 2 - 3 | 841 | 119 | ||
| XM_015443200.1 | XP_015298686.1 | 2 - 3 | 841 | 119 | ||
| XM_015443201.1 | XP_015298687.1 | 2 - 3 | 841 | 119 | ||
More Information
Set Membership: |
|

ACSF3
Genomic Map