Assay Details
| Gene Symbol: C6H5orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015451199.1 | XP_015306685.1 | 1 | 240 | 98 | |
| XM_015451200.1 | XP_015306686.1 | 1 | 240 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015451199.1, XM_015451200.1 transcripts.
Set Membership: |
|

C6H5orf38
Genomic Map