Assay Details
| Gene Symbol: IL6R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541663.2 | XP_005541720.2 | 5 - 6 | 1573 | 93 | |
| XM_015456651.1 | XP_015312137.1 | 6 - 7 | 1691 | 93 | ||
| XM_015456656.1 | XP_015312142.1 | 6 - 7 | 1722 | 93 | ||
More Information
Set Membership: |
|

IL6R
Genomic Map