Assay Details
| Gene Symbol: FCRLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454466.1 | XP_015309952.1 | 2 - 3 | 223 | 134 | |
| XM_015454469.1 | XP_015309955.1 | 4 - 5 | 287 | 134 | ||
| XM_015454478.1 | XP_015309964.1 | 2 - 3 | 223 | 134 | ||
More Information
Set Membership: |
|

FCRLB
Genomic Map