Assay Details
| Gene Symbol: CORO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577072.2 | XP_005577129.1 | 3 - 4 | 524 | 127 | |
| XM_005577073.2 | XP_005577130.1 | 3 - 4 | 517 | 127 | ||
| XM_005577074.2 | XP_005577131.1 | 3 - 4 | 504 | 127 | ||
| XM_005577075.2 | XP_005577132.1 | 2 - 3 | 340 | 127 | ||
More Information
Set Membership: |
|

CORO1B
Genomic Map