Assay Details
| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572250.2 | XP_005572307.1 | 7 - 8 | 935 | 93 | |
| XM_005572251.2 | XP_005572308.1 | 5 - 6 | 1300 | 93 | ||
More Information
Set Membership: |
|

HVCN1
Genomic Map