Assay Details
| Gene Symbol: FCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580478.2 | XP_005580535.1 | 1 - 2 | 239 | 156 | |
| XM_015436233.1 | XP_015291719.1 | 1 - 2 | 239 | 156 | ||
More Information
Set Membership: |
|

FCN1
Genomic Map