Assay Details
| Gene Symbol: FMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319420.1 | NP_001306349.1 | 6 - 7 | 856 | 138 | |
| XM_005539976.2 | XP_005540033.1 | 7 - 8 | 1920 | 138 | ||
| XM_005539977.2 | XP_005540034.1 | 6 - 7 | 1778 | 138 | ||
| GenBank mRNA | KC734485.1 | - | 6 - 7 | 856 | 138 | |
More Information
Set Membership: |
|

FMO4
Genomic Map