Assay Details
| Gene Symbol: TNFRSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545122.2 | XP_005545179.1 | 2 - 3 | 749 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF4| Gene Symbol: TNFRSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545122.2 | XP_005545179.1 | 2 - 3 | 749 | 111 | |
Set Membership: |
|