Assay Details
| Gene Symbol: STX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588192.2 | XP_005588249.2 | 1 - 2 | 233 | 60 | |
| XM_015440337.1 | XP_015295823.1 | 1 - 2 | 233 | 60 | ||
More Information
Set Membership: |
|

STX10
Genomic Map