Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287643.1 | NP_001274572.1 | 1 - 2 | 211 | 109 | |
| XM_005553125.2 | XP_005553182.1 | 1 - 2 | 320 | 109 | ||
| XM_005553126.2 | XP_005553183.1 | 1 - 2 | 320 | 109 | ||
| GenBank mRNA | AB170198.1 | - | 1 - 2 | 211 | 109 | |
More Information
Set Membership: |
|

RNF8
Genomic Map