Assay Details
| Gene Symbol: NEURL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566358.1 | XP_005566415.1 | 4 - 5 | 1671 | 106 | |
| XM_005566359.2 | XP_005566416.1 | 4 - 5 | 1432 | 106 | ||
| XM_015456644.1 | XP_015312130.1 | 4 - 5 | 1740 | 106 | ||
More Information
Set Membership: |
|

NEURL1
Genomic Map