Assay Details
| Gene Symbol: AURKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582834.2 | XP_005582891.1 | 8 - 9 | 1102 | 149 | |
| XM_005582835.2 | XP_005582892.1 | 7 - 8 | 1034 | 149 | ||
| XM_005582836.2 | XP_005582893.1 | 7 - 8 | 1026 | 149 | ||
| XM_005582837.2 | XP_005582894.1 | 7 - 8 | 1087 | 149 | ||
| XM_005582839.2 | XP_005582896.1 | 8 - 9 | 1109 | 149 | ||
| XM_015437621.1 | XP_015293107.1 | 7 - 8 | 1095 | 149 | ||
| XM_015437622.1 | XP_015293108.1 | 3 - 4 | 411 | 149 | ||
More Information
Set Membership: |
|

AURKB
Genomic Map