Assay Details
| Gene Symbol: SLC22A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546667.2 | XP_005546724.1 | 1 - 2 | 634 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A13| Gene Symbol: SLC22A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546667.2 | XP_005546724.1 | 1 - 2 | 634 | 93 | |
Set Membership: |
|