Assay Details
| Gene Symbol: ERG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548686.2 | XP_005548743.1 | 10 - 11 | 1089 | 105 | |
| XM_005548687.2 | XP_005548744.1 | 9 - 10 | 1111 | 105 | ||
| XM_005548688.2 | XP_005548745.1 | 9 - 10 | 1017 | 105 | ||
| XM_005548689.2 | XP_005548746.1 | 8 - 9 | 1036 | 105 | ||
| XM_015446856.1 | XP_015302342.1 | 9 - 10 | 1381 | 105 | ||
More Information
Set Membership: |
|

ERG
Genomic Map