Assay Details
| Gene Symbol: ARHGAP35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589672.1 | XP_005589729.1 | 1 - 2 | 3869 | 73 | |
| XM_005589673.2 | XP_005589730.1 | 1 - 2 | 3869 | 73 | ||
| XM_015441410.1 | XP_015296896.1 | 1 - 2 | 3869 | 73 | ||
More Information
Set Membership: |
|

ARHGAP35
Genomic Map