Assay Details
| Gene Symbol: SLC7A6OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592345.1 | XP_005592402.1 | 3 - 4 | 710 | 92 | |
| GenBank mRNA | AB170130.1 | - | 3 - 4 | 698 | 92 | |
More Information
Set Membership: |
|

SLC7A6OS
Genomic Map