Assay Details
| Gene Symbol: CYP3A43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319505.1 | NP_001306434.1 | 3 - 4 | 295 | 142 | |
| XM_015447122.1 | XP_015302608.1 | 3 - 4 | 577 | 142 | ||
| GenBank mRNA | DQ074796.1 | - | 3 - 4 | 295 | 142 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015447122.1 transcripts.
Set Membership: |
|

CYP3A43
Genomic Map