Assay Details
| Gene Symbol: RIPOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569341.2 | XP_005569398.1 | 21 - 22 | 3227 | 119 | |
| XM_015429966.1 | XP_015285452.1 | 20 - 21 | 3080 | 119 | ||
| XM_015429967.1 | XP_015285453.1 | 20 - 21 | 2702 | 119 | ||
| XM_015429968.1 | XP_015285454.1 | 21 - 22 | 3144 | 119 | ||
More Information
Set Membership: |
|

RIPOR3
Genomic Map