Assay Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590019.2 | XP_005590076.1 | 3 - 4 | 672 | 142 | |
| XM_005590020.2 | XP_005590077.1 | 3 - 4 | 486 | 142 | ||
| XM_005590021.2 | XP_005590078.1 | 3 - 4 | 675 | 142 | ||
More Information
Set Membership: |
|

JOSD2
Genomic Map