Assay Details
| Gene Symbol: CMTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592157.2 | XP_005592214.1 | 1 - 2 | 241 | 106 | |
| XM_015442890.1 | XP_015298376.1 | 1 - 2 | 241 | 106 | ||
| XM_015442891.1 | XP_015298377.1 | 1 - 2 | 241 | 106 | ||
More Information
Set Membership: |
|

CMTM3
Genomic Map