Assay Details
| Gene Symbol: TSPAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542228.2 | XP_005542285.1 | 3 - 4 | 329 | 157 | |
| XM_005542230.2 | XP_005542287.1 | 3 - 4 | 331 | 157 | ||
More Information
Set Membership: |
|

TSPAN2
Genomic Map