Assay Details
| Gene Symbol: NAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571290.2 | XP_005571347.1 | 5 - 6 | 1574 | 121 | |
| GenBank mRNA | AB171555.1 | - | 5 - 6 | 384 | 121 | |
More Information
Set Membership: |
|

NAB2
Genomic Map