Assay Details
| Gene Symbol: ST8SIA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560552.2 | XP_005560609.1 | 3 - 4 | 516 | 133 | |
| XM_015453423.1 | XP_015308909.1 | 2 - 3 | 2270 | 133 | ||
More Information
Set Membership: |
|

ST8SIA2
Genomic Map