Assay Details
| Gene Symbol: CD300A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584850.2 | XP_005584907.1 | 5 - 6 | 1051 | 104 | |
| XM_005584851.2 | XP_005584908.1 | 5 - 6 | 1034 | 104 | ||
| XM_005584853.2 | XP_005584910.1 | 5 - 6 | 1045 | 104 | ||
More Information
Set Membership: |
|

CD300A
Genomic Map