Assay Details
| Gene Symbol: PYCARD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442582.1 | XP_015298068.1 | 1 - 2 | 1119 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PYCARD| Gene Symbol: PYCARD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442582.1 | XP_015298068.1 | 1 - 2 | 1119 | 147 | |
Set Membership: |
|