Assay Details
| Gene Symbol: LRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563903.2 | XP_005563960.1 | 3 - 4 | 341 | 129 | |
| XM_005563904.2 | XP_005563961.1 | 2 - 3 | 284 | 129 | ||
More Information
Set Membership: |
|

LRP12
Genomic Map