Assay Details
| Gene Symbol: LHX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580886.2 | XP_005580943.1 | 4 - 5 | 600 | 101 | |
| XM_005580887.2 | XP_005580944.1 | 4 - 5 | 600 | 101 | ||
| XM_005580889.2 | XP_005580946.1 | 4 - 5 | 441 | 101 | ||
More Information
Set Membership: |
|

LHX6
Genomic Map