Assay Details
| Gene Symbol: NUAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015431474.1 | XP_015286960.1 | 1 - 2 | 1650 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NUAK1| Gene Symbol: NUAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015431474.1 | XP_015286960.1 | 1 - 2 | 1650 | 94 | |
Set Membership: |
|