Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583613.2 | XP_005583670.1 | 2 - 3 | 1224 | 79 | |
| XM_015438227.1 | XP_015293713.1 | 1 - 2 | 1272 | 79 | ||
| XM_015438228.1 | XP_015293714.1 | 1 - 2 | 1284 | 79 | ||
More Information
Set Membership: |
|

ZNF652
Genomic Map