Assay Details
| Gene Symbol: RHOBTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562834.2 | XP_005562891.1 | 2 - 3 | 725 | 96 | |
| XM_005562835.2 | XP_005562892.1 | 2 - 3 | 1657 | 96 | ||
| XM_015454580.1 | XP_015310066.1 | 2 - 3 | 291 | 96 | ||
More Information
Set Membership: |
|

RHOBTB2
Genomic Map