Assay Details
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539621.2 | XP_005539678.1 | 4 - 5 | 668 | 116 | |
| XM_005539622.2 | XP_005539679.1 | 4 - 5 | 900 | 116 | ||
More Information
Set Membership: |
|

DESI2
Genomic Map