Assay Details
| Gene Symbol: ENPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552823.2 | XP_005552880.1 | 3 - 4 | 1247 | 111 | |
| XM_005552824.2 | XP_005552881.1 | 4 - 5 | 1312 | 111 | ||
| XM_005552826.2 | XP_005552883.1 | 3 - 4 | 1140 | 111 | ||
More Information
Set Membership: |
|

ENPP5
Genomic Map