Assay Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588030.2 | XP_005588087.1 | 3 - 4 | 459 | 68 | |
| XM_005588032.2 | XP_005588089.1 | 3 - 4 | 459 | 68 | ||
More Information
Set Membership: |
|

PLPPR2
Genomic Map