Assay Details
| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588341.2 | XP_005588398.1 | 2 - 3 | 373 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM38A| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588341.2 | XP_005588398.1 | 2 - 3 | 373 | 73 | |
Set Membership: |
|